Calciumbis(dihydrogenorthophosphat) Hydrat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Calcium dihydrogen phosphate hydrate
A12123.0C
A12123.0E
A12123.36
A12123.0CEA
471
EUR
A12123.0C
A12123.0E
A12123.36
Calciumbis(dihydrogenorthophosphat) Hydrat ≥97%
Calciumbis(dihydrogenorthophosphat) Hydrat
Purity: 97%
CAS No.: 301524-28-7
Molecular Formula: Ca(H2PO4)2.xH2O
Formula Weight: 234.06(anhy)
Flash Point: None
Melting Point: ca 200° dec.
MDL No.: MFCD00010898
Formel:
H₄CaO₈P₂·H₂O Molecular Weight: 252,07 g/mol Schmelzpunkt: ∼ 200 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010898 CAS-Nummer: 301524-28-7 EINECS: 231-837-1 Merck Index: 14,1693 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...