(-)-Menthon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
L-Menthon
A13679.09
A13679.18
A13679.22
A13679.30
A13679.09EA
27
EUR
A13679.09
A13679.18
A13679.22
A13679.30
(-)-Menthon ≥97%
(-)-Menthon
Purity: 97%
CAS No.: 14073-97-3
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 72°(162°F)
Physical Form: Clear liquid
Density: 0.893
Refractive Index: 1.4500
Appearance: Colourless
Boiling Point: 208-210°
Beilstein Reference: 3648743
Merck Reference: 14,5838
Optical Rotation: -25° (neat)
MDL No.: MFCD00001634
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 208…210 °C Dichte: 0,893 Flammpunkt: 72 °C (162 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001634 CAS-Nummer: 14073-97-3 EINECS: 237-926-1 Merck Index: 14,5838 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...