5-Brom-2-furancarbonsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid
A14840.18
A14840.30
A14840.18EA
62.1
EUR
A14840.18
A14840.30
5-Brom-2-furancarbonsäure ≥99%
5-Bromo-2-furoic acid
Purity: 99%
CAS No.: 585-70-6
Molecular Formula: C5H3BrO3
Formula Weight: 190.99
Melting Point: 188-192°
Beilstein Reference: 118354
Formel:
C₅H₃BrO₃ Molecular Weight: 190,98 g/mol Schmelzpunkt: 188…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003239 CAS-Nummer: 585-70-6 EINECS: 209-559-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...