Methyl-5-nitro-2-furoat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A15226.06
A15226.14
A15226.06EA
69.1
EUR
A15226.06
A15226.14
Methyl-5-nitro-2-furoat ≥97%
Methyl-5-nitro-2-furoat
Purity: 97%
CAS No.: 1874-23-3
Molecular Formula: C6H5NO5
Formula Weight: 171.11
Melting Point: 78-82°
Beilstein Reference: 165132
Formel:
C₆H₅NO₅ Molecular Weight: 171,11 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051956 CAS-Nummer: 1874-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...