(3-Chlorpropyl)dimethylammoniumchlorid 98%
Lieferant: Thermo Scientific
Gefahr
Synonyme: 3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride
A15459.0E A15459.22 A15459.36
A15459.0EEA 410 EUR
A15459.0E A15459.22 A15459.36
(3-Chlorpropyl)dimethylammoniumchlorid 98%
3-Chloropropyldimethylammonium chloride
Purity: 98% CAS No.: 5407-04-5 Molecular Formula: Cl(CH2)3N(CH3)2.HCl Formula Weight: 158.07 Melting Point: 138-144° Beilstein Reference: 3670046
Formel: C₅H₁₃Cl₂N
Molecular Weight: 158,07 g/mol
Schmelzpunkt: 138…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012521
CAS-Nummer: 5407-04-5
EINECS: 226-467-2