4-Chlorpyridin Hydrochlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Chloropyridinium chloride
A15628.06
A15628.14
A15628.22
A15628.06EA
24.2
EUR
A15628.06
A15628.14
A15628.22
4-Chlorpyridin Hydrochlorid ≥98%
4-Chlorpyridin Hydrochlorid
Purity: 98+%
CAS No.: 7379-35-3
Molecular Formula: C5H4ClN.HCl
Formula Weight: 150.01
Flash Point: 198°(388°F)
Odour: Weak
pH: 2
Melting Point: ca 210° subl.
Beilstein Reference: 3684904
MDL No.: MFCD00012829
Formel:
C₅H₅Cl₂N Schmelzpunkt: ∼ 210 °C (Subl.) Flammpunkt: 198 °C (388 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012829 CAS-Nummer: 7379-35-3 EINECS: 230-946-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...