TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure) ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

A17754.14 A17754.22
A17754.14EA 41.4 EUR
A17754.14 A17754.22
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure) ≥99%
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)

Purity: 99% CAS No.: 29915-38-6 Molecular Formula: (HOCH2)3CNH(CH2)3SO3H Formula Weight: 243.28 Melting Point: 239-241° dec. MDL No.: MFCD00007538

Formel: C₇H₁₇NO₆S
Schmelzpunkt: 239…241 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007538
CAS-Nummer: 29915-38-6
EINECS: 249-954-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR