Aluminiumtrifluormethansulfonat 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

B20785.06 B20785.14
B20785.06EA 75.1 EUR
B20785.06 B20785.14
Aluminiumtrifluormethansulfonat 99%
Aluminiumtrifluormethansulfonat

Purity: 99% CAS No.: 74974-61-1 Molecular Formula: C3AlF9O9S3 Formula Weight: 474.20 Melting Point: ca 300° UN No.: 3261 MDL No.: MFCD00143596

Formel: C₃AlF₉O₉S₃
Molecular Weight: 474,19 g/mol
Schmelzpunkt: ∼ 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143596
CAS-Nummer: 74974-61-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR