Rubidiumtetrafluoroborat ≥98%

Lieferant: Thermo Scientific
Achtung

B20817.06 B20817.14
B20817.06EA 76.5 EUR
B20817.06 B20817.14
Rubidiumtetrafluoroborat ≥98%
Rubidium tetrafluoroborate

Purity: 98% CAS No.: 18909-68-7 Molecular Formula: BF4Rb Formula Weight: 172.27 MDL No.: MFCD00083071

Formel: BF₄Rb
Lagertemperatur: Raumtemperatur
MDL: MFCD00083071
CAS-Nummer: 18909-68-7
EINECS: 242-660-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR