2-Methyl-5-phenyl-3-furancarbonsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methyl-5-phenylfuran-3-carboxylic acid

B20988.03 B20988.06
B20988.03EA 77.6 EUR
B20988.03 B20988.06
2-Methyl-5-phenyl-3-furancarbonsäure ≥97%
2-Methyl-5-phenyl-3-furoic acid

Purity: 97% CAS No.: 108124-17-0 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 181-184° MDL No.: MFCD00221070

Formel: C₁₂H₁₀O₃
Molecular Weight: 202,21 g/mol
Schmelzpunkt: 181…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221070
CAS-Nummer: 108124-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR