3-Methylphthalsäureanhydrid 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Methylisobenzofuran-1,3-dione

B22864.06 B22864.14
B22864.06EA 116 EUR
B22864.06 B22864.14
3-Methylphthalsäureanhydrid 96%
3-Methylphthalsäureanhydrid

Purity: 96% CAS No.: 4792-30-7 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 114-116° MDL No.: MFCD00047316

Formel: C₉H₆O₃
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047316
CAS-Nummer: 4792-30-7
EINECS: 225-344-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR