α-Acetyl-γ-butyrolacton ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone
B22967.22 B22967.36
B22967.22EA 32.3 EUR
B22967.22 B22967.36
α-Acetyl-γ-butyrolacton ≥98%
α-Acetyl-γ-butyrolacton
Purity: 98+% CAS No.: 517-23-7 Molecular Formula: C6H8O3 Formula Weight: 128.13 Flash Point: 135°(275°F) Refractive Index: 1.4590 Boiling Point: 107-108°/5mm Beilstein Reference: 112676 Merck Reference: 14,83 MDL No.: MFCD00005394
Formel: C₆H₈O₃
Molecular Weight: 128.13 g/mol
Siedepunkt: 107…108 °C (5 mmHg)
Dichte: 1.190
Flammpunkt: 135 °C (275 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005394
CAS-Nummer: 517-23-7
EINECS: 208-235-2