Dichlor(methyl)phenylsilan ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Methylphenyldichlorosilane

B23999.14 B23999.22 B23999.36
B23999.14EA 27.6 EUR
B23999.14 B23999.22 B23999.36
Dichlor(methyl)phenylsilan ≥98%
Dichloro(methyl)(phenyl)silane

Purity: 98% CAS No.: 149-74-6 Molecular Formula: C7H8Cl2Si Formula Weight: 191.13 Flash Point: 82°(179°F) Refractive Index: 1.5190 Odour: Acrid Boiling Point: 205-206° Beilstein Reference: 970975 UN No.: 2437 MDL No.: MFCD00000490

Formel: C₇H₈Cl₂Si
Molecular Weight: 191,13 g/mol
Siedepunkt: 205…206 °C
Dichte: 1,173
Flammpunkt: 82 °C (179 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000490
CAS-Nummer: 149-74-6
EINECS: 205-746-2
UN: 2437
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR