2-Aminoterephthalsäure 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Aminoterephthalic Acid
B24441.06
B24441.14
B24441.22
B24441.06EA
31.4
EUR
B24441.06
B24441.14
B24441.22
2-Aminoterephthalsäure 99%
2-Aminoterephthalsäure
Purity: 99%
CAS No.: 10312-55-7
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Odour: Odourless
Melting Point:
Formel:
C₈H₇NO₄ Molecular Weight: 181,15 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134536 CAS-Nummer: 10312-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...