2-Methyl-3-furancarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methylfuran-3-carboxylic acid
B24928.06
B24928.14
B24928.06EA
46.8
EUR
B24928.06
B24928.14
2-Methyl-3-furancarbonsäure ≥98%
2-Methyl-3-furoic acid
Purity: 98%
CAS No.: 6947-94-0
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: 97-99°
Beilstein Reference: 112229
Formel:
C₆H₆O₃ Molecular Weight: 126,11 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092314 CAS-Nummer: 6947-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...