1-(4-Aminophenyl)-4-methylpiperazin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline
H26684.03
H26684.06
H26684.03EA
142
EUR
H26684.03
H26684.06
1-(4-Aminophenyl)-4-methylpiperazin ≥97%
1-(4-Aminophenyl)-4-methylpiperazin
Purity: 97%
CAS No.: 16153-81-4
Molecular Formula: C11H17N3
Formula Weight: 191.27
Melting Point: 89-94°
UN No.: 3263
MDL No.: MFCD00172703
Formel:
C₁₁H₁₇N₃ Molecular Weight: 191,28 g/mol Schmelzpunkt: 89…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172703 CAS-Nummer: 16153-81-4 EINECS: None UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...