3-Chlor-5-fluorpyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H30170.03
H30170.06
H30170.03EA
96
EUR
H30170.03
H30170.06
3-Chlor-5-fluorpyridin ≥98%
3-Chloro-5-fluoropyridine
Purity: 98%
CAS No.: 514797-99-0
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Refractive Index: 1.5015
Melting Point: 25-27°
UN No.: 1325
MDL No.: MFCD05663707
Formel:
C₅H₃ClFN Schmelzpunkt: 25…27 °C Flammpunkt: 41 °C (106 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663707 CAS-Nummer: 514797-99-0 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...