2-Brom-6-methoxypyridin ≥97%

Lieferant: Thermo Scientific
Achtung

H30593.06 H30593.14
H30593.06EA 47.2 EUR
H30593.06 H30593.14
2-Brom-6-methoxypyridin ≥97%
2-Bromo-6-methoxypyridine

Purity: 97% CAS No.: 40473-07-2 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Flash Point: 104°(219°F) Refractive Index: 1.5600 Boiling Point: 206° MDL No.: MFCD00088345

Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
Siedepunkt: 206 °C
Dichte: 1,530
Flammpunkt: 104 °C (219 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00088345
CAS-Nummer: 40473-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR