4-(Boc-amino)pyridin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Boc-Amino)pyridine

H30820.06 H30820.14
H30820.06EA 33.9 EUR
H30820.06 H30820.14
4-(Boc-amino)pyridin 95%
4-(Boc-amino)pyridin

Purity: 95% CAS No.: 98400-69-2 Molecular Formula: C10H14N2O2 Formula Weight: 194.23 Melting Point: 147-150° MDL No.: MFCD02179232

Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194,23 g/mol
Schmelzpunkt: 147…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179232
CAS-Nummer: 98400-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR