2,4,6-Trimethylpyryliumtetrafluoroborat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,4,6-Trimethylpyryliumtetrafluoroborate
L00467.06
L00467.14
L00467.06EA
49.6
EUR
L00467.06
L00467.14
2,4,6-Trimethylpyryliumtetrafluoroborat ≥98%
2,4,6-Trimethylpyrylium tetrafluoroborate
Purity: 98+%
CAS No.: 773-01-3
Molecular Formula: C8H11BF4O
Formula Weight: 209.98
Storage Temperature: 2°C to 8°C
Melting Point: 224-226°
Beilstein Reference: 3639361
MDL No.: MFCD00032098
Formel:
C₈H₁₁BF₄O Molecular Weight: 209.98 g/mol Schmelzpunkt: 224…226 °C |
MDL:
MFCD00032098 CAS-Nummer: 773-01-3 EINECS: 212-256-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...