o-Toluylsäurehydrazid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
o-Toluic acid hydrazide, 2-Methylbenzohydrazide
L00668.06
L00668.14
L00668.06EA
42
EUR
L00668.06
L00668.14
o-Toluylsäurehydrazid ≥98%
o-Toluylsäurehydrazid
Purity: 98+%
CAS No.: 7658-80-2
Molecular Formula: C8H10N2O
Formula Weight: 150.18
Melting Point: 122-124°
Beilstein Reference: 1862809
Formel:
C₈H₁₀N₂O Molecular Weight: 150.18 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014756 CAS-Nummer: 7658-80-2 EINECS: 231-623-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...