Tropinon ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

L02011.06 L02011.14 L02011.22 L02011.PG L02011.A9
L02011.06EA 33.2 EUR
L02011.06 L02011.14 L02011.22 L02011.PG L02011.A9
Tropinon ≥99%
Tropinon

Purity: 99% CAS No.: 532-24-1 Molecular Formula: C8H13NO Formula Weight: 139.20 Storage Temperature: 2°C to 8°C Flash Point: 90°(194°F) Odour: Odourless Melting Point: 40-43° Boiling Point: 227° Beilstein Reference: 2329 MDL No.: MFCD00005549

Formel: C₈H₁₃NO
Molecular Weight: 139.2 g/mol
Siedepunkt: 227 °C
Schmelzpunkt: 40…43 °C
Flammpunkt: 90 °C (194 °F)
MDL: MFCD00005549
CAS-Nummer: 532-24-1
EINECS: 208-530-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR