Tropin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
L02332.09
L02332.18
L02332.09EA
40
EUR
L02332.09
L02332.18
Tropin ≥98%
Tropin
Purity: 98%
CAS No.: 120-29-6
Molecular Formula: C8H15NO
Formula Weight: 141.22
Storage Temperature: 2°C to 8°C
Odour: Amine-like
pH: 12
Melting Point: 61-65° (dried)
Boiling Point: 232-234°
Beilstein Reference: 80188
Merck Reference: 14,9781
MDL No.: MFCD00005551
Formel:
C₈H₁₅NO Molecular Weight: 141,21 g/mol Siedepunkt: 232…234 °C Schmelzpunkt: 61…65 °C (wasserfrei) Dichte: 1,04 |
MDL:
MFCD00180667 CAS-Nummer: 120-29-6 EINECS: 204-384-2 Merck Index: 14,9781 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...