Chlor(methyl)diphenylsilan ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

L04162.09 L04162.18
L04162.09EA 36.5 EUR
L04162.09 L04162.18
Chlor(methyl)diphenylsilan ≥97%
Chlor(methyl)diphenylsilan

Purity: 97% CAS No.: 144-79-6 Molecular Formula: C13H13ClSi Formula Weight: 232.79 Flash Point: 141°(285°F) Refractive Index: 1.5740 Odour: Irritating Boiling Point: 294-295° Beilstein Reference: 2937445 UN No.: 2987 MDL No.: MFCD00000498

Formel: C₁₃H₁₃ClSi
Molecular Weight: 232.78 g/mol
Siedepunkt: 294…295 °C
Dichte: 1.104
Flammpunkt: 141 °C (285 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000498
CAS-Nummer: 144-79-6
EINECS: 205-639-0
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR