2-Furoylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L06960.18
L06960.30
L06960.18EA
56.1
EUR
L06960.18
L06960.30
2-Furoylchlorid ≥98%
2-Furoylchlorid
Purity: 98+%
CAS No.: 527-69-5
Molecular Formula: C5H3ClO2
Formula Weight: 130.53
Flash Point: 85°(185°F)
Refractive Index: 1.5310
Melting Point: -2 to 1°
Boiling Point: 173-174°
Beilstein Reference: 110144
Merck Reference: 14,4310
UN No.: 3265
MDL No.: MFCD00003228
Formel:
C₅H₃ClO₂ Molecular Weight: 130,53 g/mol Siedepunkt: 173…174 °C Schmelzpunkt: –2…+1 °C Dichte: 1,325 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003228 CAS-Nummer: 527-69-5 EINECS: 208-422-9 UN: 3265 ADR: 8,II Merck Index: 14,4310 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...