Cer(III)trifluormethansulfonat 98%

Lieferant: Thermo Scientific
Achtung

L20251.06 L20251.14
L20251.06EA 74.3 EUR
L20251.06 L20251.14
Cer(III)trifluormethansulfonat 98%
Cer(III)trifluormethansulfonat

Purity: 98% CAS No.: 76089-77-5 Molecular Formula: C3CeF9O9S3 Formula Weight: 587.33 Melting Point: 300° MDL No.: MFCD02093899

Formel: C₃CeF₉O₉S₃
Schmelzpunkt: >300 °C
Dichte: 1,7
Lagertemperatur: Raumtemperatur
MDL: MFCD02093899
CAS-Nummer: 76089-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR