1-(4-Aminophenyl)-4-methylpiperazin
Lieferant: BIONET RESEARCH
Synonyme:
4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline
10G-420S.500
10G-420S.1
10G-420S.5
10G-420S.10
BIOT10G-420S.500EA
0
EUR
BIOT10G-420S.500
BIOT10G-420S.1
BIOT10G-420S.5
BIOT10G-420S.10
1-(4-Aminophenyl)-4-methylpiperazin
1-(4-Aminophenyl)-4-methylpiperazin
Formel:
C₁₁H₁₇N₃ Molecular Weight: 191,28 g/mol |
MDL:
MFCD00172703 CAS-Nummer: 16153-81-4 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...