Methyl-3,4-dichlorbenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 3,4-dichlorobenzoate, Methyl 3,4-dichlorobenzenecarboxylate

10H-383S.500 10H-383S.1 10H-383S.5 10H-383S.10
BIOT10H-383S.500EA 0 EUR
BIOT10H-383S.500 BIOT10H-383S.1 BIOT10H-383S.5 BIOT10H-383S.10
Methyl-3,4-dichlorbenzoat
Methyl-3,4-dichlorbenzoat
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00018165
CAS-Nummer: 2905-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR