6-Phenoxy-3-formylpyridin

Lieferant: BIONET RESEARCH

Synonyme: 6-Phenoxynicotinaldehyde, 6-Phenoxypyridine-3-carboxaldehyde

10X-0989.1 10X-0989.10 10X-0989.500 10X-0989.5
BIOT10X-0989.1EA 0 EUR
BIOT10X-0989.1 BIOT10X-0989.10 BIOT10X-0989.500 BIOT10X-0989.5
6-Phenoxy-3-formylpyridin
6-Phenoxy-3-formylpyridin
Formel: C₁₂H₉NO₂
Molecular Weight: 199.21 g/mol
MDL: MFCD02681950
CAS-Nummer: 173282-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR