Methyl-3,4-difluorbenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 3,4-difluorobenzoate

12X-0841.10 12X-0841.1 12X-0841.5 12X-0841.500
BIOT12X-0841.10EA 0 EUR
BIOT12X-0841.10 BIOT12X-0841.1 BIOT12X-0841.5 BIOT12X-0841.500
Methyl-3,4-difluorbenzoat
Methyl-3,4-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD00276597
CAS-Nummer: 369-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR