3-Furoylchlorid
Lieferant: BIONET RESEARCH
7X-0706.500
7X-0706.1
7X-0706.5
7X-0706.10
BIOT7X-0706.500EA
0
EUR
BIOT7X-0706.500
BIOT7X-0706.1
BIOT7X-0706.5
BIOT7X-0706.10
3-Furoylchlorid
3-Furoylchlorid
Formel:
C₅H₃ClO₂ Molecular Weight: 130.53 g/mol |
MDL:
MFCD02681970 CAS-Nummer: 26214-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...