1-(7-Methoxy-1-benzofuran-2-yl)ethanon
Lieferant: BIONET RESEARCH
Synonyme:
1-(7-Methoxy-1-benzofuran-2-yl)-1-ethanone, 1-(7-Methoxybenzofuran-2-yl)ethanone
8P-918.500
8P-918.1
8P-918.5
BIOT8P-918.500EA
0
EUR
BIOT8P-918.500
BIOT8P-918.1
BIOT8P-918.5
1-(7-Methoxy-1-benzofuran-2-yl)ethanon
2-Acetyl-7-methoxybenzofuran
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190,2 g/mol |
MDL:
MFCD00016911 CAS-Nummer: 43071-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...