4-Fluor-3-nitrobenzonitril

Lieferant: CHEMPUR

05-0023.1 05-0023.5
CHMP05-0023.1EA 0 EUR
CHMP05-0023.1 CHMP05-0023.5
4-Fluor-3-nitrobenzonitril
4-Fluor-3-nitrobenzonitril
Formel: C₇H₃FN₂O₂
MDL: MFCD01632197
CAS-Nummer: 1009-35-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR