Chlor(dimethyl)phenylsilan

Lieferant: CHEMPUR

Synonyme: Chlorodimethylphenylsilane

BTB13912.250 BTB13912.1
CHMPBTB13912.250EA 0 EUR
CHMPBTB13912.250 MAYBBTB13912.1
Chlor(dimethyl)phenylsilan
Chlor(dimethyl)phenylsilan
Formel: C₈H₁₁ClSi
Molecular Weight: 170,71 g/mol
MDL: MFCD00000499
CAS-Nummer: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR