2-Furancarbonsäure

Lieferant: Thermo Scientific

Synonyme: Furan-2-carboxylic acid, 2-Furancarboxylic acid

CC02001.1 CC02001.10
MAYBCC02001.1EA 0 EUR
MAYBCC02001.1 MAYBCC02001.10
2-Furancarbonsäure
2-Furoic acid
Formel: C₅H₄O₃
Molecular Weight: 112,08 g/mol
MDL: MFCD00003238
CAS-Nummer: 88-14-2
EINECS: 201-803-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR