2-Furoylchlorid

Lieferant: CHEMPUR

CC02002.250 CC02002.1
CHMPCC02002.250EA 0 EUR
CHMPCC02002.250 MAYBCC02002.1
2-Furoylchlorid
2-Furoylchlorid
Formel: C₅H₃ClO₂
Molecular Weight: 130,53 g/mol
MDL: MFCD00003228
CAS-Nummer: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR