Pyridin-2-carbaldehyd

Lieferant: Thermo Scientific

Synonyme: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde

CC04004.250 CC04004.1
MAYBCC04004.250EA 0 EUR
MAYBCC04004.250 MAYBCC04004.1
Pyridin-2-carbaldehyd
Pyridin-2-carbaldehyd
Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
MDL: MFCD00006290
CAS-Nummer: 1121-60-4
EINECS: 214-333-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR