4-Fluor-2-nitrobenzonitril

Lieferant: Thermo Scientific

KM06283.250 KM06283DA
MAYBKM06283.250EA 0 EUR
MAYBKM06283.250 MAYBKM06283DA
4-Fluor-2-nitrobenzonitril
4-Fluor-2-nitrobenzonitril
Formel: C₇H₃FN₂O₂
MDL: MFCD00277447
CAS-Nummer: 80517-21-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR