Methyl-5-brom-2-furoat
Lieferant: Thermo Scientific
Synonyme:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
KM08612.1
KM08612.10
MAYBKM08612.1EA
0
EUR
MAYBKM08612.1
MAYBKM08612.10
Methyl-5-brom-2-furoat
Methyl-5-brom-2-furoat
Formel:
C₆H₅BrO₃ Molecular Weight: 205,01 g/mol |
MDL:
MFCD00092313 CAS-Nummer: 2527-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...