5-Nitro-2-furoylchlorid
Lieferant: Thermo Scientific
Synonyme:
5-Nitrofuran-2-carbonyl chloride
RF02312.1
RF02312.10
RF02312.50
MAYBRF02312.1EA
0
EUR
MAYBRF02312.1
MAYBRF02312.10
MAYBRF02312.50
5-Nitro-2-furoylchlorid
5-Nitro-2-furoylchlorid
Formel:
C₅H₂ClNO₄ Molecular Weight: 175,53 g/mol |
MDL:
MFCD00039564 CAS-Nummer: 25084-14-4 EINECS: 246-607-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...