Methyl-4-(trifluormethyl)benzoat
Lieferant: CHEMPUR
Synonyme:
Methyl 4-(trifluoromethyl)benzoate
RF03471.250
RF03471.1
CHMPRF03471.250EA
0
EUR
CHMPRF03471.250
MAYBRF03471.1
Methyl-4-(trifluormethyl)benzoat
Methyl-4-(trifluormethyl)benzoat
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol |
MDL:
MFCD00042324 CAS-Nummer: 2967-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...