Ethyl-2-furoat

Lieferant: Thermo Scientific

SB00846.1 SB00846.10 SB00846.50
MAYBSB00846.1EA 0 EUR
MAYBSB00846.1 MAYBSB00846.10 MAYBSB00846.50
Ethyl-2-furoat
Ethyl-2-furoat
Formel: C₇H₈O₃
MDL: MFCD00003237
CAS-Nummer: 614-99-3
EINECS: 210-404-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR