Ethyl-3-furoat
Lieferant: Thermo Scientific
Synonyme: Ethyl furan-3-carboxylate, Ethyl 3-Furoate, Ethyl 3-Furancarboxylate, 3-Furancarboxylic acid ethyl ester, 3-Furoic acid ethyl ester
SB01306.1 SB01306.10
MAYBSB01306.1EA 0 EUR
MAYBSB01306.1 MAYBSB01306.10
Ethyl-3-furoat
Ethyl-3-furoat
Formel: C₇H₈O₃
Molecular Weight: 140,14 g/mol
MDL: MFCD00005348
CAS-Nummer: 614-98-2
EINECS: 210-403-5