4-Picolylchlorid Hydrochlorid
Lieferant: Thermo Scientific
Synonyme:
4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride
SB01930.1
SB01930.10
SB01930.50
MAYBSB01930.1EA
0
EUR
MAYBSB01930.1
MAYBSB01930.10
MAYBSB01930.50
4-Picolylchlorid Hydrochlorid
4-Picolylchlorid Hydrochlorid
Formel:
C₆H₇Cl₂N Molecular Weight: 164.03 g/mol |
MDL:
MFCD00012826 CAS-Nummer: 1822-51-1 EINECS: 217-350-7 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...