Methyl-2,6-difluorbenzoat

Lieferant: Thermo Scientific

Synonyme: Methyl 2,6-difluorobenzoate

TL00222.1 TL00222.10 TL00222.50
MAYBTL00222.1EA 0 EUR
MAYBTL00222.1 MAYBTL00222.10 MAYBTL00222.50
Methyl-2,6-difluorbenzoat
Methyl-2,6-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD00051778
CAS-Nummer: 13671-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR