Methyl-3-(trifluormethyl)benzoat
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-(trifluoromethyl)benzoate
TL00281.1
TL00281.10
TL00281.50
MAYBTL00281.1EA
0
EUR
MAYBTL00281.1
MAYBTL00281.10
MAYBTL00281.50
Methyl-3-(trifluormethyl)benzoat
Methyl-3-(trifluormethyl)benzoat
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol |
MDL:
MFCD00043543 CAS-Nummer: 2557-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...