Methyl-2-(trifluormethoxy)benzoat

Lieferant: Thermo Scientific

TL00419.1 TL00419.10
MAYBTL00419.1EA 0 EUR
MAYBTL00419.1 MAYBTL00419.10
Methyl-2-(trifluormethoxy)benzoat
Methyl-2-(trifluormethoxy)benzoat
Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
MDL: MFCD00052336
CAS-Nummer: 148437-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR