2-Chlor-4-nitrobenzonitril

Lieferant: Thermo Scientific

XAX00005.1 XAX00005EA
MAYBXAX00005.1EA 0 EUR
MAYBXAX00005.1 MAYBXAX00005EA
2-Chlor-4-nitrobenzonitril
2-Chlor-4-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
MDL: MFCD00052203
CAS-Nummer: 28163-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR