1-Boc-3-hydroxypiperidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-butyl 3-hydroxypiperidine-1-carboxylate

AM-1751-5G AM-1751-25G AM-1751-1G
COBBAM-1751-5GEA 25.9 EUR
COBBAM-1751-5G COBBAM-1751-25G COBBAM-1751-1G
1-Boc-3-hydroxypiperidin
1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201,27 g/mol
MDL: MFCD02093938
CAS-Nummer: 85275-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR