2,6-Difluor-3-nitrobenzonitril
Lieferant: COMBI-BLOCKS
AN-1061-5G
AN-1061-25G
AN-1061-1G
AN-1061-100G
COBBAN-1061-5GEA
49.8
EUR
COBBAN-1061-5G
COBBAN-1061-25G
COBBAN-1061-1G
COBBAN-1061-100G
2,6-Difluor-3-nitrobenzonitril
2,6-Difluor-3-nitrobenzonitril
Formel:
C₇H₂F₂N₂O₂ Molecular Weight: 184.1 g/mol |
MDL:
MFCD00042254 CAS-Nummer: 143879-77-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...